Novus Biologicals
Manufacturer Code:NBP297508G
Catalog # NB102396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human IFT122 fragment. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas) intraflagellar transport protein 122 homolog SPGWD repeat-containing protein 140 WD repeat domain 10 WD repeat-containing protein 10 WDR10WDR10p WDR140CED; Intraflagellar transport protein 122 homolog; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140
Gene Aliases: CED; CED1; IFT122; SPG; WDR10; WDR10p; WDR140
UniProt ID: (Human) Q9HBG6
Entrez Gene ID: (Human) 55764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.