Invitrogen
Manufacturer Code:PA589158
Catalog # PIPA589158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-240 of human IFRD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12-O-tetradecanoylphorbol-13-acetate-induced sequence 7; Interferon-related developmental regulator 1; IRPR; Nerve growth factor-inducible protein PC4; pheochromocytoma cell-4; TIS7 protein; TPA induced sequence 7; TPA-induced sequence 7
Gene Aliases: Ifnl; IFRD1; PC4; TIS7
UniProt ID: (Human) O00458, (Mouse) P19182, (Rat) P20695
Entrez Gene ID: (Human) 3475, (Mouse) 15982, (Rat) 29596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.