Invitrogen
Manufacturer Code:MA530679
Catalog # PIMA530679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 74 |
Immunogen | Recombit Mouse IFNGR1/CD119 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; IFNGR1 IFNGR AVP type 2 CD119 antigen CDw119 IFN-gamma receptor 1 IFN-gamma-R1 antiviral protein type 2 immune interferon receptor 1 interferon-gamma receptor alpha chain; INF-g receptor; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1
Gene Aliases: CD119; Ifgr; IFN-gammaR; Ifngr; Ifngr1; Nktar
UniProt ID: (Mouse) P15261
Entrez Gene ID: (Mouse) 15979
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.