Invitrogen
Manufacturer Code:PA599641
Catalog # PIPA599641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IFN-gammaRalpha corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antiviral protein type 2 AVP type 2 CD119 CD119 antigen CDw119 Ifgr IFN-gamma receptor 1 IFN-gammaR IFN-gamma-R1 IFN-gamma-R-alpha Ifngr Ifngr1 IMD27A IMD27B immune interferon receptor 1 INF-g receptor interferon gamma receptor 1 Interferon gamma receptor alpha-chain interferon-gamma receptor 1 interferon-gamma receptor alpha chain Nktar; antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; INF-g receptor; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain
Gene Aliases: CD119; Ifgr; IFN-gammaR; IFNGR; IFNGR1; IMD27A; IMD27B; Nktar
UniProt ID: (Human) P15260, (Mouse) P15261
Entrez Gene ID: (Human) 3459, (Mouse) 15979
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.