Invitrogen
Manufacturer Code:PA586322
Catalog # PIPA586322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 430-480 of Human IFN-gammaRalpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; INF-g receptor; Interferon gamma receptor 1; Interferon gamma receptor 1 IFN-gamma receptor 1 IFN-gamma-R1 CDw119 CD119 IFNGR1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain
Gene Aliases: CD119; Ifgr; IFN-gammaR; IFNGR; IFNGR1; IMD27A; IMD27B; Nktar
UniProt ID: (Human) P15260, (Mouse) P15261
Entrez Gene ID: (Human) 3459, (Mouse) 15979, (Rat) 116465
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.