Abnova
Manufacturer Code:H00003459K
Catalog # 89-208-118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human IFNGR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human IFNGR1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CD119 FLJ45734 IFNGR; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor alpha chain
Gene Aliases: CD119; IFNGR; IFNGR1; IMD27A; IMD27B
UniProt ID: (Human) P15260
Entrez Gene ID: (Human) 3459
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.