Abnova
Manufacturer Code:MAB14562
Catalog # 89-034-645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | IFNG/466 |
Immunogen | Recombinant protein corresponding to full length human IFNG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IFG; IFI; IFN-gamma; Immune interferon; Interferon gamma; interferon, gamma
Gene Aliases: IFG; IFI; IFNG
UniProt ID: (Human) P01579
Entrez Gene ID: (Human) 3458
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.