Abnova
Manufacturer Code:MAB12440
Catalog # 89-934-343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B11 |
Immunogen | Native purified bovine IFNG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at &minus20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against bovine IFNG.
For Research Use Only
Protein Aliases: BoIFNG; IFN-gamma; Interferon gamma; interferon gamma type 2; interferon-gamma precursor
Gene Aliases: IFNG
UniProt ID: (Bovine) P07353
Entrez Gene ID: (Bovine) 281237
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.