Abnova
Manufacturer Code:PAB12999
Catalog # 89-116-254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region 17 amino acids of human IFNB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of IFNB1.
For Research Use Only
Protein Aliases: Fibroblast interferon; IFB; IFF; IFN-beta; IFNB; Interferon beta; interferon, beta 1, fibroblast; interferon-beta; MGC96956
Gene Aliases: IFB; IFF; IFN-beta; IFNB; IFNB1
UniProt ID: (Human) P01574
Entrez Gene ID: (Human) 3456
Molecular Function:
cytokine
interferon superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.