Invitrogen
Manufacturer Code:PA528303
Catalog # PIPA528303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 118 and 431 of Human IFNAR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: human interferon alpha/beta receptor; IFN-alpha/beta receptor 2; IFN-R IFN-alpha-REC IFNABR IFNARB IFN-R-2 IFN-alpha binding protein IFN-alpha/beta receptor 2 human interferon alpha/beta receptor interferon alpha binding protein interferon alpha/beta receptor 2 interferon-alpha/beta receptor beta chain type I interferon receptor 2; IFN-R-2; interferon (alpha, beta and omega) receptor 2; Interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; Type I interferon receptor 2
Gene Aliases: IFN-alpha-REC; IFN-R; IFNABR; IFNAR2; IFNARB; IMD45
UniProt ID: (Human) P48551
Entrez Gene ID: (Human) 3455
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.