Proteintech
Manufacturer Code:105221AP150UL
Catalog # 50-172-6375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IFNAR2 Fusion Protein Ag0191 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: human interferon alpha/beta receptor; IFN alpha REC IFN alpha/beta receptor 2 IFN R IFN R 2 IFNABR IFNAR2 IFNARB Type I interferon receptor 2; IFN-alpha/beta receptor 2; IFN-R-2; interferon (alpha, beta and omega) receptor 2; Interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; Type I interferon receptor 2
Gene Aliases: IFN-alpha-REC; IFN-R; IFNABR; IFNAR2; IFNARB; IMD45
UniProt ID: (Human) P48551
Entrez Gene ID: (Human) 3455
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.