Invitrogen
Manufacturer Code:12594580
Catalog # 50-112-2358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MAR1-5A3 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFN-R-1; INF-a receptor; Interferon alpha/beta receptor 1; interferon-alpha/beta receptor 1; Type I interferon receptor 1
Gene Aliases: CD118; Ifar; Ifnar; Ifnar1; Ifrc; Infar
UniProt ID: (Mouse) P33896
Entrez Gene ID: (Mouse) 15975
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.