Invitrogen
Manufacturer Code:MA524850
Catalog # PIMA524850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1G10 |
Immunogen | Recombit IFNAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-type antiviral protein; beta-type antiviral protein; CRF2-1; Cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFN-R-1; INF-a receptor; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; Interferon alpha/beta receptor 1; Interferon alpha/beta receptor 1 IFN-R-1 IFN-alpha/beta receptor 1 Cytokine receptor class-II member 1 Cytokine receptor family 2 member 1 CRF2-1 Type I interferon receptor 1 IFNAR1 IFNAR IFNR1 IFN R1 IFN-alpha/betaRalphaIFN-alpha/betaR1 IFN alpha/betaRalpha IFNalpha/betaR1 IFNalpha/beta-RalphaIFNalpha/beta R1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; Type I interferon receptor 1
Gene Aliases: AVP; CD118; Ifar; IFN-alpha-REC; IFNAR; IFNAR1; IFNBR; IFRC; Infar
UniProt ID: (Human) P17181, (Mouse) P33896
Entrez Gene ID: (Human) 3454, (Rat) 288264, (Mouse) 15975
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.