Invitrogen
Manufacturer Code:PA547436
Catalog # PIPA547436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IFN-alpha/beta R1 Glu27-Thr429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AVP IFN-alpha-REC IFNAR IFNBR IFRC CRF2-1 IFN-R-1 IFN-alpha/beta receptor 1 alpha-type antiviral protein beta-type antiviral protein cytokine receptor class-II member 1 cytokine receptor family 2 member 1 interferon alpha/beta receptor 1 interferon-alpha/beta receptor alpha chain interferon-beta receptor 1 type I interferon receptor 1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFN-R-1; INF-a receptor; Interferon alpha/beta receptor 1; interferon-alpha/beta receptor 1; Type I interferon receptor 1
Gene Aliases: CD118; Ifar; Ifnar; Ifnar1; Ifrc; Infar
UniProt ID: (Mouse) P33896
Entrez Gene ID: (Mouse) 15975
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.