Invitrogen
Manufacturer Code:PA538503
Catalog # PIPA538503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human Interferon-alpha/beta Receptor alpha chain |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-type antiviral protein; AVP IFN-alpha-REC IFNAR IFNBR IFRC CRF2-1 IFN-R-1 IFN-alpha/beta receptor 1 alpha-type antiviral protein beta-type antiviral protein cytokine receptor class-II member 1 cytokine receptor family 2 member 1 interferon alpha/beta receptor 1 interferon-alpha/beta receptor alpha chain interferon-beta receptor 1 type I interferon receptor 1; beta-type antiviral protein; CRF2-1; Cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; IFN-alpha/beta receptor 1; IFN-R-1; interferon (alpha, beta and omega) receptor 1; Interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; Type I interferon receptor 1
Gene Aliases: AVP; IFN-alpha-REC; IFNAR; IFNAR1; IFNBR; IFRC
UniProt ID: (Human) P17181
Entrez Gene ID: (Human) 3454
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.