Abnova
Manufacturer Code:PAB9607
Catalog # 89-134-270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S535/S539 of human IFNAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of IFNAR1.
For Research Use Only
Protein Aliases: alpha-type antiviral protein; AVP; beta-type antiviral protein; CRF2-1; Cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; IFN-alpha-REC; IFN-alpha/beta receptor 1; IFN-R-1; IFNAR; IFNBR; IFRC; interferon (alpha, beta and omega) receptor 1; Interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; Type I interferon receptor 1
Gene Aliases: AVP; IFN-alpha-REC; IFNAR; IFNAR1; IFNBR; IFRC
UniProt ID: (Human) P17181
Entrez Gene ID: (Human) 3454
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.