Novus Biologicals
Manufacturer Code:NBP190223
Catalog # NBP190223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ASPSAGFPMDFNVTLRLRAELGALHSAWVTMPWFQHYRNVTVGPPENIEVTPGEGSLIIRFSSPFDIADTSTAFFCYYVHYWEKGGIQQVKGPFRSNSISLDNLKPSRVYCLQVQAQLLWNKSNIFRVGHLSNIS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AF-1; AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1 IFGR2 IFN-gamma receptor 2 IFN-gamma-R2 interferon gamma receptor 2 interferon gamma receptor 2 (interferon gamma transducer 1) interferon gamma receptor accessory factor-1 interferon gamma receptor beta chain Interferon gamma transducer 1; IFN-gamma receptor 2; IFN-gamma-R-beta; IFN-gamma-R2; Interferon gamma receptor 2; Interferon gamma receptor accessory factor 1; interferon gamma receptor accessory factor-1; interferon gamma receptor beta chain; Interferon gamma receptor beta-chain; Interferon gamma transducer 1
Gene Aliases: AF-1; IFGR2; IFNGR2; IFNGT1; IMD28
UniProt ID: (Human) P38484
Entrez Gene ID: (Human) 3460
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.