R&D Systems
Manufacturer Code:AF773SP
Catalog # AF773SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R2 Ala22-Gln247 Accession # P38484 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AF-1; AF-1 AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1 IFGR2 IFN-gamma receptor 2 IFNgammaR2 IFN-gamma-R2 IFN-gR2 IFNGR2 IFNGT1 interferon gamma receptor 2 interferon gamma receptor 2 (interferon gamma transducer 1) interferon gamma receptor accessory factor-1 interferon gamma receptor beta chain Interferon gamma transducer 1; IFN-gamma receptor 2; IFN-gamma-R-beta; IFN-gamma-R2; Interferon gamma receptor 2; Interferon gamma receptor accessory factor 1; interferon gamma receptor accessory factor-1; interferon gamma receptor beta chain; Interferon gamma receptor beta-chain; Interferon gamma transducer 1
Gene Aliases: AF-1; IFGR2; IFNGR2; IFNGT1; IMD28
UniProt ID: (Human) P38484
Entrez Gene ID: (Human) 3460
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.