R&D Systems
Manufacturer Code:FAB673F
Catalog # FAB673F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 92101 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1 Glu18-Gly245 Accession # P15260 |
Conjugate | Fluorescein |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antiviral protein, type 2; AVP type 2 CD119 CD119 antigen FLJ45734 IFN-gamma receptor 1 IFNgammaR1 IFN-gamma-R1 IFNGR IFN-gR1 IFNGR1 immune interferon receptor 1 interferon gamma receptor 1 interferon-gamma receptor alpha chain type 2; AVP, type 2; CD119; CD119 antigen; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor alpha chain
Gene Aliases: CD119; IFNGR; IFNGR1; IMD27A; IMD27B
UniProt ID: (Human) P15260
Entrez Gene ID: (Human) 3459
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.