R&D Systems
Manufacturer Code:AF1026
Catalog # AF1026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IFN-gamma R1/CD119 Ala26-Asp253 Accession # Q91Y85 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: antiviral protein, type 2; AVP type 2 CD119 CD119 antigen FLJ45734 IFN-gamma receptor 1 IFNgammaR1 IFN-gamma-R1 IFNGR IFN-gR1 IFNGR1 immune interferon receptor 1 interferon gamma receptor 1 interferon-gamma receptor alpha chain type 2; AVP, type 2; CD119; CD119 antigen; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor alpha chain
Gene Aliases: CD119; IFNGR; IFNGR1; IMD27A; IMD27B
UniProt ID: (Human) P15260
Entrez Gene ID: (Human) 3459
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.