R&D Systems
Manufacturer Code:MAB4851500
Catalog # MAB4851500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Chinese hamster |
Class | Monoclonal |
Type | Antibody |
Clone | H22 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse IFN-gamma Accession # P01580 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: IFG IFI IFNG IFNgamma IFN-gamma Immune interferon interferon gamma interferon gamma; IFN-gamma; Immune interferon; Interferon gamma; interferon, gamma
Gene Aliases: IFG; IFI; IFNG
UniProt ID: (Human) P01579
Entrez Gene ID: (Human) 3458
Molecular Function:
cytokine
interferon superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.