R&D Systems
Manufacturer Code:214701
Catalog # 214701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MMHB-16 |
Immunogen | Human interferon beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | After receipt the product may be stored at -20degreesC for short-term use (< 6 months). For long-term storage we recommend storing the product at -70degreesC or below for retention of full activity. When thawing the contents of the tube should be a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fibroblast interferon; Fibroblast interferon IFB IFBIFNB IFF IFNB IFNB1 IFNbeta IFN-beta interferon beta interferon beta 1 fibroblast MGC96956; IFN-beta; Interferon beta; interferon, beta 1, fibroblast; interferon-beta
Gene Aliases: IFB; IFF; IFN-beta; IFNB; IFNB1
UniProt ID: (Human) P01574
Entrez Gene ID: (Human) 3456
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.