R&D Systems
Manufacturer Code:MAB8234100
Catalog # MAB8234100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1176D |
Immunogen | Human embryonic kidney cell line HEK293-derived mouse IFN-beta Ile22-Asn182 Accession # P01575 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Fibroblast interferon; Fibroblast interferon IFB IFBIFNB IFF IFNB IFNB1 IFNbeta IFN-beta interferon beta interferon beta 1 fibroblast MGC96956; IFN-beta; Interferon beta; interferon, beta 1, fibroblast; interferon-beta
Gene Aliases: IFB; IFF; IFN-beta; IFNB; IFNB1
UniProt ID: (Human) P01574
Entrez Gene ID: (Human) 3456
Molecular Function:
cytokine
interferon superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.