Novus Biologicals
Manufacturer Code:NBP131603
Catalog # NBP131603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human interferon alpha 8. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha-8; IFN-alpha-8 IFN-alphaB interferon alpha type 201 interferon alpha-8 interferon alpha-B' Interferon alpha-B Interferon alpha-B2 interferon alpha 8 LeIF B; interferon alpha type 201; Interferon alpha-8; Interferon alpha-B; interferon alpha-B'; Interferon alpha-B2; interferon, alpha 8; LeIF B
Gene Aliases: IFN-alphaB; IFNA8
UniProt ID: (Human) P32881
Entrez Gene ID: (Human) 3445
Molecular Function:
cytokine
interferon superfamily
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.