Invitrogen
Manufacturer Code:221001
Catalog # PI221001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RMMA-1 |
Immunogen | mouse interferon alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -80° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha 1b IFN-alpha-1/13 interferon alpha 1b interferon alpha-1/13 interferon alpha-D interferon-alpha1 leIF D; IFN-alpha-1; interferon alpha family gene 1; Interferon alpha-1
Gene Aliases: Ifa1; Ifna1
UniProt ID: (Mouse) P01572
Entrez Gene ID: (Mouse) 15962
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.