Invitrogen
Manufacturer Code:271001
Catalog # PI271001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | K9 |
Immunogen | Porcine Interferon Alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha-6; IFN-alpha-6 interferon alpha 1 interferon-alpha-6; interferon-alpha; interferon-alpha-6
Gene Aliases: IFN-ALPHA-1; IFN1@
Entrez Gene ID: (Pig) 397686
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.