Invitrogen
Manufacturer Code:710299
Catalog # PI710299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 9HCLC |
Immunogen | Recombit protein corresponding to amino acids 24189 of human Interferon-alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: IFN-alpha 1b; IFN-alpha 1b IFN-alpha-1/13 interferon alpha 1b interferon alpha-1/13 interferon alpha-D interferon-alpha1 leIF D; IFN-alpha-1/13; interferon alpha 1b; Interferon alpha-1/13; Interferon alpha-D; interferon, alpha 1; interferon-alpha1; LeIF D
Gene Aliases: IFL; IFN; IFN-ALPHA; IFN-alphaD; IFNA1; IFNA13; IFNA@
UniProt ID: (Human) P01562
Entrez Gene ID: (Human) 3439
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.