Invitrogen
Manufacturer Code:PA184928
Catalog # PIPA184928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit chicken Interferon alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IFN alpha; IFN-alpha IFN-gamma IFNA IFNA1 IFNA2 IFNA6; interferon alpha; Interferon type A1/A2; interferon type A3
Gene Aliases: IFN-alpha; IFN-gamma; IFNA; IFNA1; IFNA2; IFNA6
UniProt ID: (Chicken) P42165
Entrez Gene ID: (Chicken) 396398
Molecular Function: cytokine interferon superfamily signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.