Invitrogen
Manufacturer Code:PA524534
Catalog # PIPA524534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 55-83 amino acids from the central region of human IFITM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bone-restricted ifitm-like protein; bone-restricted interferon induced transmembrane protein-like protein; Bone-restricted interferon-induced transmembrane protein-like protein; BRIL; BRIL Hrmp1 fragilis4 bone-restricted interferon induced transmembrane protein-like protein bone-restricted interferon-induced transmembrane protein-like protein interferon-induced transmembrane protein 5; Dispanin subfamily A member 1; DSPA1; Fragilis family member 4; fragilis4; haemopoiesis related membrane protein 1; Interferon-induced transmembrane protein 5
Gene Aliases: 1110003J06Rik; AW213665; BRIL; DSPA1; Fragilis4; Hrmp1; Hrtm1; IFITM5; OI5
UniProt ID: (Human) A6NNB3, (Mouse) O88728
Entrez Gene ID: (Human) 387733, (Mouse) 73835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.