Invitrogen
Manufacturer Code:MA537720
Catalog # PIMA537720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1082CT14.1.3.1 |
Immunogen | KLH conjugated synthetic peptide between 1-133 amino acids from the N-terminal region of human IFITM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1110004C05Rik 1-8U Cd225 Cdw217 Dispanin subfamily A member 2b DSPA2b Fgls Fragilis Fragilis 3 Fragilis protein Ifitm3 ifitm-like protein 1 interferon induced transmembrane protein 3 interferon-induced transmembrane protein 3 interferon-inducible protein 15 Interferon-inducible protein 1-8U interferon-inducible protein variant 10 IP15 mil-1 Mouse ifitm-like protein 1 rat8; Dispanin subfamily A member 2b; DSPA2b; Interferon-induced transmembrane protein 3; Interferon-inducible protein 1-8U
Gene Aliases: 1-8U; DSPA2b; IFITM3; IP15
UniProt ID: (Human) Q01628
Entrez Gene ID: (Human) 10410
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.