Abnova
Manufacturer Code:PAB2360
Catalog # 89-125-305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human IFITM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of IFITM3.
For Research Use Only
Protein Aliases: 1-8U; Dispanin subfamily A member 2b; DSPA2b; Interferon-induced transmembrane protein 3; Interferon-inducible protein 1-8U; IP15
Gene Aliases: 1-8U; DSPA2b; IFITM3; IP15
UniProt ID: (Human) Q01628
Entrez Gene ID: (Human) 10410
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.