Invitrogen
Manufacturer Code:PA141257
Catalog # 01-675-603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 1-50 of mouse fragilis. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dispanin subfamily A member 2b; DSPA2b; Fragilis 3; Fragilis protein; ifitm-like protein 1; Interferon-induced transmembrane protein 3; Interferon-inducible protein 15; interferon-inducible protein variant 10; Mil-1; Mouse ifitm-like protein 1
Gene Aliases: 1110004C05Rik; Cd225; Cdw217; DSPA2b; Fgls; Ifitm3; IP15; mil-1; rat8
UniProt ID: (Rat) P26376, (Mouse) Q9CQW9
Entrez Gene ID: (Rat) 361673, (Mouse) 66141
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.