Invitrogen
Manufacturer Code:PA520989
Catalog # PIPA520989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the center of human IFITM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9-27 CD225 IFI17 LEU13 interferon-induced protein 17 interferon-induced transmembrane protein 1 interferon-inducible protein 9-27 leu-13 antigen; CD225; Dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; ifitm-like protein 2; interferon induced transmembrane protein 2 like; Interferon-induced protein 17; Interferon-induced transmembrane protein 1; Interferon-inducible protein 9-27; Leu-13 antigen; Mil-2; Mouse ifitm-like protein 2
Gene Aliases: 1110036C17Rik; 9-27; CD225; DSPA2a; IFI17; IFITM1; LEU13; Mil-2; Mil2
UniProt ID: (Human) P13164, (Mouse) Q9D103
Entrez Gene ID: (Human) 8519, (Rat) 293618, (Mouse) 68713
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.