Novus Biologicals
Manufacturer Code:NBP271005AF594
Catalog # NP27105A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G8 |
Immunogen | Full length recombinant protein of human IFIT1 (NP_000899) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: G10P1C56 GARG-16 IFI56IFI-56 IFI-56K IFIT-1 IFNAI1ISG56 Interferon alpha-inducible protein (MW 56kD) Interferon-induced 56 kDa protein interferon-induced protein 56 interferon-induced protein with tetratricopeptide repeats 1 p56 RNM561; IFI-56K; IFIT-1; interferon, alpha-inducible protein (MW 56kD); Interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-inducible mRNA 561
Gene Aliases: C56; G10P1; IFI-56; IFI-56K; IFI56; IFIT-1; IFIT1; IFNAI1; ISG56; P56; RNM561
UniProt ID: (Human) P09914
Entrez Gene ID: (Human) 3434
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.