Novus Biologicals
Manufacturer Code:NBP30443220UL
Catalog # NB094999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human IFI6 (NP_002029.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAM14C G1P3interferon alpha-inducible protein 6 6-16 Ifi-6-16 IFI616interferon alpha-inducible protein (clone IFI-6-16) interferon alpha-inducible protein 6 interferon alpha-inducible protein clone IFI-6-16 Interferon-induced protein 6-16; Ifi-6-16; Interferon alpha-inducible protein 6; interferon, alpha-inducible protein 6; interferon, alpha-inducible protein clone IFI-6-16; Interferon-induced protein 6-16
Gene Aliases: 6-16; FAM14C; G1P3; IFI-6-16; IFI6; IFI616
UniProt ID: (Human) P09912
Entrez Gene ID: (Human) 2537
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.