Invitrogen
Manufacturer Code:PA576462
Catalog # PIPA576462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human IFI16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gamma-interferon-inducible protein 16; Gamma-interferon-inducible protein Ifi-16 Interferon-inducible myeloid differentiation transcriptional activator IFI16 IFNGIP1; Ifi-16; interferon, gamma-inducible protein 16; interferon-gamma induced protein IFI 16; Interferon-inducible myeloid differentiation transcriptional activator
Gene Aliases: IFI16; IFNGIP1; PYHIN2
UniProt ID: (Human) Q16666
Entrez Gene ID: (Human) 3428
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.