Invitrogen
Manufacturer Code:MA523317
Catalog # PIMA523317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H4 |
Immunogen | IDNK (NP&lowbar001001551 41 a.a. &bsim 141 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCRG2 KIAA1824 WDR22; glucokinase-like protein; Gluconate kinase; gluconokinase-like protein; idnK, gluconokinase homolog; Probable gluconokinase
Gene Aliases: bA522I20.2; C9orf103; hGntK; IDNK
UniProt ID: (Human) Q5T6J7
Entrez Gene ID: (Human) 414328
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.