Novus Biologicals
Manufacturer Code:NBP297025AF700
Catalog # NB112274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant IDI2 (Q9BXS1 Met1-Val227). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: diphosphate dimethylallyl diphosphate isomerase 2; diphosphate dimethylallyl diphosphate isomerase 2 EC 5.3.3.2 IPP isomerase 2 IPPI2isopentenyl-diphosphate Delta-isomerase 2 Isopentenyl pyrophosphate isomerase 2 isopentenyl-diphosphate delta isomerase 2; IPP isomerase 2; Isopentenyl pyrophosphate isomerase 2; Isopentenyl-diphosphate delta-isomerase 2
Gene Aliases: IDI2; IPPI2
UniProt ID: (Human) Q9BXS1
Entrez Gene ID: (Human) 91734
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.