Invitrogen
Manufacturer Code:PA521720
Catalog # PIPA521720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 173 of IDI1 (Uniprot IDNo.Q13907) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IPP isomerase 1; IPP1 IPPI1 IPP isomerase 1 isopentenyl diphosphate dimethylallyl diphosphate isomerase 1 isopentenyl pyrophosphate isomerase 1 isopentenyl-diphosphate Delta-isomerase 1; isopentenyl diphosphate dimethylallyl diphosphate isomerase 1; Isopentenyl pyrophosphate isomerase 1; Isopentenyl-diphosphate Delta-isomerase 1
Gene Aliases: IDI1; IPP1; IPPI1
UniProt ID: (Human) Q13907
Entrez Gene ID: (Human) 3422
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.