Proteintech
Manufacturer Code:111662AP150UL
Catalog # 50-172-6931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IDI1 Fusion Protein Ag1636 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IDI1 IPP isomerase 1 IPP1 IPPI1; IPP isomerase 1; IPPI1; isopentenyl diphosphate dimethylallyl diphosphate isomerase 1; Isopentenyl pyrophosphate isomerase 1; Isopentenyl-diphosphate Delta-isomerase 1
Gene Aliases: 4832416K17Rik; IDI1; IPP1; IPPI1
UniProt ID: (Human) Q13907, (Rat) O35760, (Mouse) P58044
Entrez Gene ID: (Human) 3422, (Rat) 89784, (Mouse) 319554
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.