Invitrogen
Manufacturer Code:MA519176
Catalog # PIMA519176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A2-1D3 |
Immunogen | IDH3G (AAH00933 1 a.a. &bsim 393 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H-IDHG IDH-gamma NAD (H)-specific isocitrate dehydrogenase gamma subunit NAD(+)-specific ICDH subunit gamma NAD+-specific ICDH isocitrate dehydrogenase &lsqbNAD] subunit gamma mitochondrial isocitrate dehydrogenase NAD(+)-specific mitochondrial gamma subunit isocitric dehydrogenase subunit gamma; IDH-gamma; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 gamma; Isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; Isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-specific ICDH
Gene Aliases: H-IDHG; IDH3G
UniProt ID: (Human) P51553
Entrez Gene ID: (Human) 3421
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.