Invitrogen
Manufacturer Code:PA585385
Catalog # PIPA585385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human IDH3G. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H-IDHG antibody IDH3G antibody NAD (H)-specific isocitrate dehydrogenase gamma subunit antibody NAD+-specific ICDH antibody isocitrate dehydrogenase NAD(+)-specific mitochondrial gamma subunit antibody isocitrate dehydrogenase &lsqbNAD] subunit gamma mitochondrial antibody NAD(+)-specific ICDH subunit gamma antibody IDH-gamma antibody isocitric dehydrogenase subunit gamma antibody isocitrate dehydrogenase 3 (NAD+) gamma antibody; IDH-gamma; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 gamma; Isocitrate dehydrogenase [NAD] subunit gamma 1, mitochondrial; Isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; Isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-specific ICDH
Gene Aliases: H-IDHG; IDH3G
UniProt ID: (Human) P51553, (Mouse) P70404
Entrez Gene ID: (Human) 3421, (Mouse) 15929
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.