Abnova
Manufacturer Code:PAB19676
Catalog # 89-121-202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 33-46 near N-terminus of human IDH3B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of IDH3B.
For Research Use Only
Protein Aliases: FLJ11043; H-IDHB; isocitrate dehydrogenase 3 (NAD+) beta; Isocitrate dehydrogenase [NAD] subunit beta, mitochondrial; Isocitric dehydrogenase subunit beta; MGC903; NAD(+)-specific ICDH subunit beta
Gene Aliases: IDH3B; RP46
UniProt ID: (Human) O43837
Entrez Gene ID: (Human) 3420
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.