Invitrogen
Manufacturer Code:MA517271
Catalog # PIMA517271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT673 |
Immunogen | Recombit fragment corresponding to a region within amino acids 56 and 345 of IDH2 (Uniprot IDNo.P48735) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D2HGA2 ICD-M IDH IDHM IDP IDPM mNADP-IDH NADP(+)-specific ICDH isocitrate dehydrogenase &lsqbNADP] mitochondrial oxalosuccinate decarboxylase; ICD-M; IDH; IDP; isocitrate dehydrogenase 2 (NADP+), mitochondrial; Isocitrate dehydrogenase [NADP], mitochondrial; NADP(+)-specific ICDH; NADP+-specific isocitrate dehydrogenase; Oxalosuccinate decarboxylase
Gene Aliases: D2HGA2; E430004F23; ICD-M; IDH; Idh-2; IDH2; IDHM; IDP; IDPM; mNADP-IDH
UniProt ID: (Human) P48735, (Mouse) P54071, (Rat) P56574
Entrez Gene ID: (Human) 3418, (Mouse) 269951, (Rat) 361596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.