Novus Biologicals
Manufacturer Code:NBP243572
Catalog # NBP243572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 673 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human IDH2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D2HGA2 EC 1.1.1.42 ICD-M IDH IDHM IDP IDPM isocitrate dehydrogenase [NADP] mitochondrial isocitrate dehydrogenase 2 (NADP+) mitochondrial mNADP-IDH NADP(+)-specific ICDH Oxalosuccinate decarboxylase; ICD-M; IDH; IDP; isocitrate dehydrogenase 2 (NADP+), mitochondrial; Isocitrate dehydrogenase [NADP], mitochondrial; NADP(+)-specific ICDH; Oxalosuccinate decarboxylase
Gene Aliases: D2HGA2; ICD-M; IDH; IDH2; IDHM; IDP; IDPM; mNADP-IDH
UniProt ID: (Human) P48735
Entrez Gene ID: (Human) 3418
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.