Proteintech
Manufacturer Code:159321AP150UL
Catalog # 50-559-295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IDH2 Fusion Protein Ag8779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ICD M IDH IDH2 IDHM IDP IDPM mNADP IDH NADP(+) specific ICDH Oxalosuccinate decarboxylase; ICD-M; IDH; IDP; isocitrate dehydrogenase 2 (NADP+), mitochondrial; Isocitrate dehydrogenase [NADP], mitochondrial; NADP(+)-specific ICDH; NADP+-specific isocitrate dehydrogenase; Oxalosuccinate decarboxylase
Gene Aliases: D2HGA2; E430004F23; ICD-M; IDH; Idh-2; IDH2; IDHM; IDP; IDPM; mNADP-IDH
UniProt ID: (Human) P48735, (Mouse) P54071, (Rat) P56574
Entrez Gene ID: (Human) 3418, (Mouse) 269951, (Rat) 361596
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.