Invitrogen
Manufacturer Code:PA570042
Catalog # PIPA570042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human IDH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: corneal epithelial crystallin; Cytosolic NADP-isocitrate dehydrogenase; epididymis luminal protein 216; epididymis secretory protein Li 26; IDH; IDH1 isocitrate dehydrogenase 1 (NADP+) soluble IDCD IDH IDP IDPC PICD NADP+-specific ICDH NADP-dependent isocitrate dehydrogenase cytosolic NADP-dependent isocitrate dehydrogenase peroxisomal OTTHUMP00000206464 OTTHUMP00000206465 OTTHUMP00000206466 oxalosuccinate decarboxylase; IDP; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase 1, soluble; Isocitrate dehydrogenase [NADP] cytoplasmic; NADP(+)-specific ICDH; NADP-cICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; Oxalosuccinate decarboxylase
Gene Aliases: AI314845; AI788952; E030024J03Rik; HEL-216; HEL-S-26; ICDH; Id-1; IDCD; IDH; Idh-1; IDH1; IDP; IDPC; NADP-CICDH; PICD
UniProt ID: (Human) O75874, (Bovine) Q9XSG3, (Sheep) Q6XUZ5, (Mouse) Q3UAV7, (Rat) P41562
Entrez Gene ID: (Horse) 100066416, (Human) 3417, (Rabbit) 100328559, (Dog) 478889, (Bovine) 281235, (Sheep) 443257, (Mouse) 15926, (Guinea pig) 100729107, (Rat) 24479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.