Abnova
Manufacturer Code:PAB19472
Catalog # 89-120-997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of <i>Saccharomyces cerevisiae</i> (S288c) IDH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of IDH1.
For Research Use Only
Protein Aliases: isocitrate dehydrogenase (NAD(+)) IDH1; Isocitrate dehydrogenase [NAD] subunit 1, mitochondrial; Isocitric dehydrogenase; NAD(+)-specific ICDH
Gene Aliases: IDH1; N2690; YNL037C
UniProt ID: (Baker's yeast) P28834
Entrez Gene ID: (Baker's yeast) 855691
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.