OriGene
Manufacturer Code:CF811776
Catalog # 50-167-0839
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 800-1019 of human IDE produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abeta-degrading protease; Insulin protease; Insulin-degrading enzyme; Insulinase; INSULYSIN
Gene Aliases: IDE; INSDEGM; INSULYSIN
UniProt ID: (Human) P14735, (Rat) P35559
Entrez Gene ID: (Human) 3416, (Rat) 25700
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.