Invitrogen
Manufacturer Code:MA527757
Catalog # PIMA527757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT143 |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of human IDE. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Abeta-degrading protease; IDE INSULYSIN insulin degrading enzyme; Insulin protease; Insulin-degrading enzyme; Insulinase; Insulysin
Gene Aliases: 1300012G03Rik; 4833415K22Rik; AA675336; AI507533; IDE; INSDEGM; INSULYSIN
UniProt ID: (Human) P14735, (Rat) P35559
Entrez Gene ID: (Human) 3416, (Rat) 25700, (Mouse) 15925
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.